Skip to Content
Merck
All Photos(1)

Key Documents

Z122882

Supelco

Sigma-Aldrich® TLC Plates

PEI-cellulose, matrix

Sign Into View Organizational & Contract Pricing

Select a Size

25 EA
€307.00

€307.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 EA
€307.00

About This Item

UNSPSC Code:
41115711
eCl@ss:
32110504
NACRES:
NC.07

€307.00


Check Cart for Availability

Request a Bulk Order

material

PEI-cellulose, matrix
polyester support

feature

binder Organic Polymer
fluorescent indicator: no

packaging

pkg of 25 sheets

manufacturer/tradename

Sigma-Aldrich

technique(s)

thin layer chromatography (TLC): suitable

L × W

20 cm × 20 cm , PEI cellulose

layer thickness

80-120 μm

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Thin-layer chromatography (TLC) plates for adsorption, partition, and ion-exchange techniques. Six media choices with glass, polyester, and aluminum backings. Silica-based adsorbent layers in Sigma-Aldrich TLC plates incorporate a polymeric binder; highly purified silica gel plates incorporate a gypsum/polymer binder.
  • Silica gel - for weakly to strongly polar separations, pore size 60 Å
  • High purity silica gel - acid washed for separation of aflatoxins
  • Cellulose - for partition chromatography, (SigmaCell Type 100 Cellulose)
  • Cellulose PEI - for separation of weak anions (amino acids, peptides) PEI plates may turn slightly yellow, affecting separation. Pre-develop plates in distilled water to minimize yellow color.
  • Chiral silica - for separation of optically active isomers by ligand exchange, pore size 60Å

Application

Sigma-Aldrich® TLC Plates was used for TLC analysis during GTPase activity assay[1] and lipid extraction/analysis.[2]

Legal Information

SIGMA-ALDRICH is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Frédéric Picard-Jean et al.
PloS one, 13(3), e0193804-e0193804 (2018-03-31)
The 5' RNA cap structure (m7GpppRNA) is a key feature of eukaryotic mRNAs with important roles in stability, splicing, polyadenylation, mRNA export, and translation. Higher eukaryotes can further modify this minimal cap structure with the addition of a methyl group
Heiko Runz et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 22(5), 1679-1689 (2002-03-07)
Generation of amyloid-beta (Abeta) from the amyloid precursor protein (APP) requires proteolytic cleavage by two proteases, beta- and gamma-secretase. Several lines of evidence suggest a role for cholesterol on secretase activities, although the responsible cellular mechanisms remain unclear. Here we
Chuang-Rung Chang et al.
The Journal of biological chemistry, 282(30), 21583-21587 (2007-06-08)
Mitochondria in cells comprise a tubulovesicular reticulum shaped by dynamic fission and fusion events. The multimeric dynamin-like GTPase Drp1 is a critical protein mediating mitochondrial division. It harbors multiple motifs including GTP-binding, middle, and GTPase effector (GED) domains that are

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service