Key Documents
Z108359
Leather gloves
size XL
Select a Size
1 PKG
€107.00
€107.00
Check Cart for Availability
Select a Size
Change View
1 PKG
€107.00
About This Item
UNSPSC Code:
46181504
NACRES:
NB.35
€107.00
Check Cart for Availability
material
leather
size
XL
sterility
non-sterile
feature
latex free
powder-free
reusable
packaging
pack of 1 pair
manufacturer/tradename
Sigma-Aldrich
General description
Constructed of specially treated leather that stays supple even at temperature extremes of -70 to 150°C. Remarkably durable, moisture, heat, and flame resistant. Provided with an elastic cinch for a no-slip fit.
Choose from one of the most recent versions:
Certificates of Analysis (COA)