Documents
Z107425
Sigma-Aldrich
Aldrich® septum-inlet adapter with stopcock and side-arm
Plain, joint: ST/NS 19/22, glass stopcock
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
material
glass stopcock
Joint
joint: ST/NS19/22
Related Categories
General description
Septum-inlets require septum Z565695 or Z565679 except where noted.
Linkage
See the Threaded Ground Glass Tutorial to see how threaded joints work.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC