WHA68097003
Whatman® alumina Anodisc filter discs 0.2 μm pore size
transparent, hydrophilic, PP support ring, 100 ea, 13 mm diam
Synonym(s):
Whatman filter, Z694983, filter membranes, membrane filters
About This Item
Recommended Products
product name
Whatman® Anodisc inorganic filter membrane, unsupported, diam. 13 mm, pore size 0.02 μm, autoclavable
material
alumina based membrane cap
sterility
non-sterile
feature
autoclavable
packaging
pkg of 100 ea
manufacturer/tradename
Whatman 6809-7003
Whatman Article No. 28420419 (US reference)
diam.
13 mm
pore size
0.02 μm
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
The Anopore inorganic membrane (Anodisc) is well suited for a wide range of laboratory filtration applications. This novel material has a precise, nondeformable honeycomb pore structure with no lateral crossovers between individual pores, that filters at precisely the stated cut-off, allowing no larger sized particles to pass through the membrane. The Anopore inorganic membrane is composed of a high purity alumina matrix that is manufactured electrochemically. The membrane also exhibits low protein binding, has minimal autofluorescence, is nontoxic, and supports cellular growth.
The precise pore structure and narrow pore size distribution of the Anopore membrane ensure a high level of particle removal efficiency. Microorganisms and particulate material are captured on the surface of the membrane for subsequent analysis by light or electron microscopy. When wet, the membrane is virtually transparent, which means that retained particles do not need to be transferred to another surface before microscopic examination.
The membrane is hydrophilic and is compatible with most solvents and aqueous material. No monomers, plasticizers, adhesives, surfactants or wetting agents are used in the manufacturing process, which eliminates sample contamination and ensures low protein binding and minimal loss of sample.
Application
- HPLC mobile phase filtration and degassing
- Ultra cleaning of solvents
- Gravimetric analysis
- Liposome extrusion
- Scanning electron microscopy studies
- Bacterial analysis by epifluorescence light microscopy
- Micrometer and nanometer filtration
- Metal nanorods formation
Features and Benefits
- Virtually transparent when wet
- Minimal autofluorescence
- Hydrophilic with wide solvent compatibility
- Non-toxic for cellular growth
Other Notes
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Which document(s) contains shelf-life or expiration date information for a given product?
If available for a given product, the recommended re-test date or the expiration date can be found on the Certificate of Analysis.
How do I get lot-specific information or a Certificate of Analysis?
The lot specific COA document can be found by entering the lot number above under the "Documents" section.
How do I find price and availability?
There are several ways to find pricing and availability for our products. Once you log onto our website, you will find the price and availability displayed on the product detail page. You can contact any of our Customer Sales and Service offices to receive a quote. USA customers: 1-800-325-3010 or view local office numbers.
What is the Department of Transportation shipping information for this product?
Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.
What orientation should Product WHA68097003, Whatman® Anodisc inorganic filter membrane, be used in?
The 0.02 μm size Whatman® Anodisc inorganic filter membrane actually has 2 pore sizes. It is 0.02 μm on the top and 0.2 μm on the bottom. They are packaged with the 0.02 μm on the top and are used in the same orientation as they are packaged.
My question is not addressed here, how can I contact Technical Service for assistance?
Ask a Scientist here.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service