WHA231100
Whatman® drain disc
diam. 47 mm
Synonym(s):
Whatman filter, Whatman filters, Z741493
About This Item
Recommended Products
material
polyethylene housing
sterility
non-sterile
packaging
pkg of 100 ea
manufacturer/tradename
Whatman 231100
Whatman Article No. 28418861 (US reference)
diam.
47 mm
Looking for similar products? Visit Product Comparison Guide
General description
Applications
- General laboratory microfiltration
- Quality control and sterility testing
- Removal of particulates from HPLC solvents
- Tissue culture media filtration
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service