WHA10380204
Whatman® quadrant folded cellulose fiber filter papers
Grade 41 FF, diam. (110 mm), pack of 500 ea
All Photos(1)
About This Item
Recommended Products
material
cellulose
feature
Grade 41 FF
Nominal Ash Content: 0.007%
packaging
pack of 500 ea
manufacturer/tradename
Cytiva
diam.
(110 mm)
thickness
(215 μm)
Related Categories
General description
Quadrant folded filter paper saves the user valuable time and provides ease of use when undertaking repetitive or multiple analyses.
- Quadrant folded filter paper saves the user valuable time.
- Quadrant folded filter paper provides ease of use when undertaking repetitive or multiple analyses.
Features and Benefits
- Quadrant folded filter paper saves the user valuable time.
- Quadrant folded filter paper provides ease of use when undertaking repetitive or multiple analyses.
Legal Information
Whatman is a registered trademark of Cytiva