Skip to Content
Merck
All Photos(1)

Key Documents

W530155

Sigma-Aldrich

Coconut oil

virgin, Certified organic (NOP)

Synonym(s):

Coconut oil from Cocos nucifera, Coconut fat, Copra oil

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12164502
grade:
Certified organic (NOP)
Halal
food allergen:
tree nuts
Pricing and availability is not currently available.

grade

Certified organic (NOP)
Halal

origin

Philippines origin

mp

23-27 °C

density

0.903 g/mL at 25 °C

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

tree nuts

Organoleptic

coconut

Looking for similar products? Visit Product Comparison Guide

General description

Virgin coconut oil (VCO) is obtained from the fresh mature kernel of Cocos nucifera.[1][2] It is mainly composed of lauric acid, which is responsible for most of its physiological and metabolic properties.[3] VCO shows better antioxidant capacity when compared to refined, bleached and deodorized coconut oil due to the presence of higher amount of phenolic compounds such as ferulic acid and p-coumaric acid.[4]

Legal Information

Certified organic by MOSA

Storage Class Code

10 - Combustible liquids

WGK

awg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Health benefits of virgin coconut oil
Carandang EV
Indian Coconut Journal, 38(9), 8-8 (2008)
Antioxidant capacity and phenolic acids of virgin coconut oil.
Marina AM, et al.
International Journal of Food Sciences and Nutrition, 60(sup2), 114-123 (2009)
Virgin coconut oil: emerging functional food oil.
Marina AM, et al.
Trends in Food Science & Technology, 20(10), 481-487 (2009)
Chemical properties of virgin coconut oil.
Marina AM, et al.
Journal of the American Oil Chemists' Society, 86(4), 301-307 (2009)
M Hollmann et al.
Journal of dairy science, 96(3), 1769-1781 (2013-01-15)
Dietary coconut oil (CNO) can reduce dry matter intake (DMI), enteric methane (eCH(4)) emissions, and milk fat yield of lactating cows. The goals of this research were to examine responses to different CNO concentrations during the habituation period (34-d) and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service