All Photos(1)
About This Item
Linear Formula:
CH3CH=C(CH3)CO2CH3
CAS Number:
Molecular Weight:
114.14
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.624
grade
Kosher
Assay
≥99%
refractive index
n20/D 1.436 (lit.)
bp
137-138 °C/758 mmHg (lit.)
density
0.95 g/mL at 25 °C (lit.)
SMILES string
COC(=O)\C(C)=C\C
Signal Word
Warning
Hazard Statements
Hazard Classifications
Flam. Liq. 3
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
95.0 °F - closed cup
Flash Point(C)
35 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service