Skip to Content
Merck
All Photos(1)

Key Documents

W457533

Sigma-Aldrich

Diisoamyl disulfide

≥98%

Synonym(s):

3-methyl-1-(3-methylbutyldisulfanyl)butane

Sign Into View Organizational & Contract Pricing

Select a Size

W457533-SAMPLE
€50.30
100 G
€271.00
1 KG
€1,020.00

€50.30


Check Cart for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
W457533-SAMPLE
€50.30
100 G
€271.00
1 KG
€1,020.00

About This Item

Empirical Formula (Hill Notation):
C10H22S2
CAS Number:
Molecular Weight:
206.41
FEMA Number:
4575
MDL number:
UNSPSC Code:
12164502
NACRES:
NA.21
grade:
Halal
biological source:
synthetic
food allergen:
no known allergens

€50.30


Check Cart for Availability

Request a Bulk OrderRequest more information

biological source

synthetic

Quality Level

grade

Halal

reg. compliance

FDA 21 CFR 117

Assay

≥98%

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

onion; sweet

SMILES string

S(SCCC(C)C)CCC(C)C

InChI

1S/C10H22S2/c1-9(2)5-7-11-12-8-6-10(3)4/h9-10H,5-8H2,1-4H3

InChI key

MPYGLNNTOXLWOB-UHFFFAOYSA-N

Related Categories

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service