Skip to Content
Merck
All Photos(2)

Key Documents

W431105

Sigma-Aldrich

Methyl cyclohexadiene

mixture of isomers, stabilized, ≥98%

Synonym(s):

Lemon hexadiene

Sign Into View Organizational & Contract Pricing

Select a Size

W431105-SAMPLE-K
€155.00
100 G
€311.00
500 G
€731.00

€155.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
W431105-SAMPLE-K
€155.00
100 G
€311.00
500 G
€731.00

About This Item

Empirical Formula (Hill Notation):
C7H10
CAS Number:
Molecular Weight:
94.15
FEMA Number:
4311
UNSPSC Code:
12164502
NACRES:
NA.21
grade:
Halal
Kosher
food allergen:
no known allergens

€155.00


Check Cart for Availability

Request a Bulk Order

grade

Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
FDA 21 CFR 117

Assay

≥98%

contains

synthetic α-Tocopherol as stabilizer

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

lemon; lime; earthy

storage temp.

2-8°C

SMILES string

C1CC=CC=C1C

InChI

1S/C7H10/c1-7-5-3-2-4-6-7/h2-3,5H,4,6H2,1H3

InChI key

QMFJIJFIHIDENY-UHFFFAOYSA-N

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

40.1 °F

Flash Point(C)

4.5 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. What is the ratio (ballpark is fine) between the isomers CAS 30640-46-1 and CAS 1888-90-0?

    1 answer
    1. The Certificate of Analysis provides the sum of the isomers as determined by GC. The ratio or quantity of each is not reported. No further information is available. See the link below to review the CofA:
      https://www.sigmaaldrich.com/certificates/COFA/W4/W431105/W431105-BULK-K____SHBN3322__.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service