Skip to Content
Merck
All Photos(1)

Key Documents

W415201

Sigma-Aldrich

Isopropenyl acetate

99%

Synonym(s):

1-Methylvinyl acetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2C(CH3)=CH2
CAS Number:
Molecular Weight:
100.12
FEMA Number:
4152
Beilstein:
1280347
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.822

biological source

synthetic

Assay

99%

refractive index

n20/D 1.401 (lit.)

bp

94 °C (lit.)

density

0.922 g/mL at 20 °C
0.909 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Organoleptic

berry

SMILES string

CC(=C)OC(C)=O

InChI

1S/C5H8O2/c1-4(2)7-5(3)6/h1H2,2-3H3

InChI key

HETCEOQFVDFGSY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

66.2 °F - closed cup

Flash Point(C)

19 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service