Skip to Content
Merck
All Photos(1)

Key Documents

W405702

Sigma-Aldrich

3-Methyl-2,4-nonanedione

≥97%, FG

Sign Into View Organizational & Contract Pricing

Select a Size

W405702-SAMPLE-K
€47.10
25 G
€93.80
100 G
€282.00

€47.10


Check Cart for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
W405702-SAMPLE-K
€47.10
25 G
€93.80
100 G
€282.00

About This Item

Empirical Formula (Hill Notation):
C10H18O2
CAS Number:
Molecular Weight:
170.25
FEMA Number:
4057
MDL number:
UNSPSC Code:
12164502
Flavis number:
07.184
NACRES:
NA.21
grade:
FG
Kosher
food allergen:
no known allergens

€47.10


Check Cart for Availability

Request a Bulk OrderRequest more information

grade

FG
Kosher

Quality Level

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117

Assay

≥97%

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

fatty; green; vegetable

SMILES string

O=C(C(C)C(=O)C)CCCCC

InChI

1S/C10H18O2/c1-4-5-6-7-10(12)8(2)9(3)11/h8H,4-7H2,1-3H3

InChI key

BGVBGAIWXAXBLP-UHFFFAOYSA-N

Related Categories

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.
Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service