Skip to Content
Merck
All Photos(1)

Documents

W401404

Sigma-Aldrich

2-Phenylethyl isothiocyanate

FG

Synonym(s):

Phenethyl isothiocyanate, 2-Phenylethyl isothiocyanate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2CH2NCS
CAS Number:
Molecular Weight:
163.24
FEMA Number:
4014
Beilstein:
2084162
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
12.193
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002

refractive index

n20/D 1.5888 (lit.)

bp

139-140 °C/11 mmHg (lit.)

density

1.094 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

green; sulfurous

SMILES string

S=C=NCCc1ccccc1

InChI

1S/C9H9NS/c11-8-10-7-6-9-4-2-1-3-5-9/h1-5H,6-7H2

InChI key

IZJDOKYDEWTZSO-UHFFFAOYSA-N

Gene Information

human ... CYP1A2(1544)

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

2-Phenylethyl isothiocyanate (2-PE ITC) is a bioactive isothiocyanate mainly found in Brassica vegetables. It is produced by the enzymatic hydrolysis of 2-phenylethyl glucosinolate. 2-PE ITC is the main compound responsible for the nematicidal effect of Brassica root tissues against (Pratylenchus neglectus Filipjev), a root-lesion nematode. This ability makes it a promising candidate for the development of soil fumigants.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reduced susceptibility of Brassica napus to Pratylenchus neglectus in plants with elevated root levels of 2-phenylethyl glucosinolate.
Potter MJ, et al.
Journal of Nematology, 31(3), 291-291 (1999)
In vitro inhibition of soil microorganisms by 2?phenylethyl isothiocyanate.
Smith BJ & Kirkegaard JA
Plant Pathology, 51(5), 585-593 (2002)
Evaluation of Antibacterial Activity of 3?Butenyl, 4?Pentenyl, 2?Phenylethyl, and Benzyl Isothiocyanate in Brassica Vegetables
Jang M, et al.
Journal of Food Science, 75(7) (2010)
Antibacterial activity and synergistic effect between watercress extracts, 2?phenylethyl isothiocyanate and antibiotics against 11 isolates of Escherichia coli from clinical and animal source.
Freitas E, et al.
Letters in Applied Microbiology, 57(4), 266-273 (2013)
Breeze E Cavell et al.
Journal of natural products, 75(6), 1051-1057 (2012-05-23)
Phenethyl isothiocyanate (1) is a natural dietary phytochemical with cytostatic, cytotoxic, and antitumor activity. The effects of 1 were investigated on the activity of mTOR, a kinase that enhances the translation of many RNAs encoding proteins critical for cancer cell

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service