Skip to Content
Merck
All Photos(1)

Documents

W396303

Sigma-Aldrich

2,3,6-Trimethylphenol

≥97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3C6H2OH
CAS Number:
Molecular Weight:
136.19
FEMA Number:
3963
Beilstein:
2042207
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
4.085

biological source

synthetic

Assay

≥97%

mp

59-62 °C (lit.)

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

SMILES string

Cc1ccc(C)c(O)c1C

InChI

1S/C9H12O/c1-6-4-5-7(2)9(10)8(6)3/h4-5,10H,1-3H3

InChI key

QQOMQLYQAXGHSU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.

Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Noriaki Nagai et al.
Biological & pharmaceutical bulletin, 38(7), 1063-1069 (2015-07-03)
Hypercalcemia is often observed in postmenopausal women as well as in patients with primary hyperparathyroidism or malignant tumors. In this study, we investigated the relationship between calcium ion (Ca(2+)) levels in lacrimal fluid and the rate of corneal wound healing
Yoshinori Kakutani et al.
Journal of cellular biochemistry, 116(7), 1325-1333 (2015-03-05)
Vascular calcification is a clinically significant component of atherosclerosis and arises from chronic vascular inflammation. Oncostatin M (OSM) derived from plaque macrophages may contribute to the development of atherosclerotic calcification. Here, we investigated the stimulatory effects of OSM on osteoblastic
Toshihiro Kushibiki et al.
Scientific reports, 5, 13114-13114 (2015-08-19)
In photodynamic therapy (PDT), cells are impregnated with a photosensitizing agent that is activated by light irradiation, thereby photochemically generating reactive oxygen species (ROS). The amounts of ROS produced depends on the PDT dose and the nature of the photosensitizer.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service