Skip to Content
Merck
All Photos(1)

Key Documents

W396211

Sigma-Aldrich

3,3,5-Trimethylcyclohexanol

≥80%

Synonym(s):

Homomenthol

Sign Into View Organizational & Contract Pricing

Select a Size

W396211-SAMPLE-K
€45.50
100 G
€63.70
1 KG
€137.00

€45.50


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
W396211-SAMPLE-K
€45.50
100 G
€63.70
1 KG
€137.00

About This Item

Empirical Formula (Hill Notation):
C9H18O
CAS Number:
Molecular Weight:
142.24
FEMA Number:
3962
MDL number:
UNSPSC Code:
12352104
Flavis number:
02.209
NACRES:
NA.05
grade:
Fragrance grade
Kosher
food allergen:
no known allergens

€45.50


Check Cart for Availability

Request a Bulk Order

grade

Fragrance grade
Kosher

Quality Level

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
FDA 21 CFR 117

Assay

≥80%

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

musty; cooling; minty; spicy

SMILES string

OC1CC(CC(C)(C1)C)C

InChI

1S/C9H18O/c1-7-4-8(10)6-9(2,3)5-7/h7-8,10H,4-6H2,1-3H3

InChI key

BRRVXFOKWJKTGG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service