Skip to Content
Merck
All Photos(1)

Key Documents

W392103

Sigma-Aldrich

2,4-Hexadienoic acid

≥99%, FCC

Synonym(s):

Sorbic acid, α-trans-γ-trans-Sorbic acid, (E,E)-1,3-Pentadiene-1-carboxylic acid, 2,4-Hexadienoic acid, 2E,4E-Hexadienoic acid, trans,trans-2,4-Hexadienoic acid, C6:2n-2,4, NSC 35405, NSC 49103, NSC 50268

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH=CHCH=CHCOOH
CAS Number:
Molecular Weight:
112.13
FEMA Number:
3921
Beilstein:
1098547
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
8.085
NACRES:
NA.21
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Agency

meets purity specifications of JECFA

reg. compliance

FCC
FDA 21 CFR 166.110
FDA 21 CFR 172.872
FDA 21 CFR 177.2260
FDA 21 CFR 181.23
FDA 21 CFR 182.3089

vapor pressure

0.01 mmHg ( 20 °C)

Assay

≥99%

form

powder

mp

132-135 °C (lit.)

solubility

ethanol: 10%, clear, colorless to slightly yellow

density

1.2 g/cm3 at 20 °C

cation traces

As: ≤3.0 ppm
Cd: ≤1.0 ppm
Hg: ≤1.0 ppm
Pb: ≤2.0 ppm

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

odorless

SMILES string

C\C=C\C=C\C(O)=O

InChI

1S/C6H8O2/c1-2-3-4-5-6(7)8/h2-5H,1H3,(H,7,8)/b3-2+,5-4+

InChI key

WSWCOQWTEOXDQX-MQQKCMAXSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

260.6 °F - closed cup

Flash Point(C)

127 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrea Toschi et al.
Molecules (Basel, Switzerland), 25(18) (2020-09-24)
Bioactive compounds, such as organic acids (OA) and nature-identical compounds (NIC), can exert a role in the protection of intestinal mucosa functionality due to their biological properties. The aim of this study was to understand the role of 2 OA
Ya-Nan Zhang et al.
Journal of hazardous materials, 358, 216-221 (2018-07-11)
Pharmaceuticals are a group of ubiquitous emerging pollutants, many of which have been shown to undergo efficient photolysis in the environment. Photochemically produced reactive intermediates (PPRIs) sensitized by the pharmaceuticals in sunlit natural waters may induce photodegradation of coexisting compounds.
Alfredo Montaño et al.
Food microbiology, 34(1), 7-11 (2013-03-19)
The aim of this research was to ascertain the lactic acid bacteria responsible for the degradation of ascorbic acid and/or potassium sorbate, isolated from packed green olives where these additives had diminished. A total of 14 isolates were recovered from
Heidy M W den Besten et al.
Applied and environmental microbiology, 78(23), 8477-8480 (2012-09-25)
Population heterogeneity complicates the predictability of the outgrowth kinetics of individual spores. Flow cytometry sorting and monitoring of the germination and outgrowth of single dormant spores allowed the quantification of acid-induced spore population heterogeneity at pH 5.5 and in the
Takashi Ohtsuki et al.
Analytica chimica acta, 734, 54-61 (2012-06-19)
An analytical method using solvent extraction and quantitative proton nuclear magnetic resonance (qHNMR) spectroscopy was applied and validated for the absolute quantification of sorbic acid (SA) in processed foods. The proposed method showed good linearity. The recoveries for samples spiked

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service