Skip to Content
Merck
All Photos(1)

Key Documents

W372609

Sigma-Aldrich

DL-Phenylalanine

99%, FCC

Synonym(s):

(±)-2-Amino-3-phenylpropionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2CH(NH2)COOH
CAS Number:
Molecular Weight:
165.19
FEMA Number:
3726
Beilstein:
3198807
EC Number:
Council of Europe no.:
10488
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
17.017
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

reg. compliance

FCC

Assay

99%

mp

266-267 °C (dec.) (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

odorless

SMILES string

NC(Cc1ccccc1)C(O)=O

InChI

1S/C9H11NO2/c10-8(9(11)12)6-7-4-2-1-3-5-7/h1-5,8H,6,10H2,(H,11,12)

InChI key

COLNVLDHVKWLRT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Imre W K Kouw et al.
The Journal of clinical endocrinology and metabolism, 100(8), 3103-3111 (2015-06-04)
The progressive loss of muscle mass with aging is accelerated in type 2 diabetes patients. It has been suggested that this is attributed to a blunted muscle protein synthetic response to food intake. The objective of the study was to
Katarzyna M Romek et al.
Proceedings of the National Academy of Sciences of the United States of America, 112(27), 8296-8301 (2015-06-25)
Tramadol, previously only known as a synthetic analgesic, has now been found in the bark and wood of roots of the African medicinal tree Nauclea latifolia. At present, no direct evidence is available as to the biosynthetic pathway of its
Raja R Pillai et al.
The Journal of nutrition, 145(5), 954-959 (2015-03-13)
Lysine requirements of well-nourished children from developing regions have been found to be similar to those of children from developed regions (33.5 mg · kg⁻¹ · d⁻¹). However, intestinal parasites have been shown to increase lysine requirements in undernourished adults, and it is not
Sohei Nakayama et al.
Cancer research, 74(20), 5891-5902 (2014-08-29)
The discovery of somatic mutations in EGFR and development of EGFR tyrosine kinase inhibitors (TKI) have revolutionized treatment for lung cancer. However, resistance to TKIs emerges in almost all patients and currently no effective treatment is available. Here, we show
Rachel Schwartz-Narbonne et al.
Scientific reports, 5, 9791-9791 (2015-06-10)
Understanding woolly mammoth ecology is key to understanding Pleistocene community dynamics and evaluating the roles of human hunting and climate change in late Quaternary megafaunal extinctions. Previous isotopic studies of mammoths' diet and physiology have been hampered by the 'mammoth

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service