Skip to Content
Merck
All Photos(1)

Key Documents

W372005

Sigma-Aldrich

cis-2-Nonen-1-ol

≥97%, stabilized, FG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5CH=CHCH2OH
CAS Number:
Molecular Weight:
142.24
FEMA Number:
3720
EC Number:
Council of Europe no.:
10292
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
2.112
NACRES:
NA.21
grade:
FG
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 872/2012

Assay

≥97%

contains

α-tocopherol, synthetic as stabilizer

refractive index

n20/D 1.45 (lit.)

bp

96 °C/10 mmHg (lit.)

density

0.844 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

cucumber; fatty; green

SMILES string

[H]\C(CO)=C(/[H])CCCCCC

InChI

1S/C9H18O/c1-2-3-4-5-6-7-8-9-10/h7-8,10H,2-6,9H2,1H3/b8-7-

InChI key

NSSALFVIQPAIQK-FPLPWBNLSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

203.0 °F - closed cup

Flash Point(C)

95.00 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service