Skip to Content
Merck
All Photos(1)

Key Documents

W369802

Sigma-Aldrich

trans-Isoeugenyl benzyl ether

≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2OC6H3-2-(OCH3)-4-CH=CHCH3
CAS Number:
Molecular Weight:
254.32
FEMA Number:
3698
Council of Europe no.:
522
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:

biological source

synthetic

grade

Halal
Kosher

Assay

≥98%

mp

59-63 °C (lit.)

Documentation

see Safety & Documentation for available documents

Organoleptic

spicy; floral; sweet

SMILES string

COc1cc(\C=C\C)ccc1OCc2ccccc2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service