Skip to Content
Merck
All Photos(1)

Key Documents

W354503

Sigma-Aldrich

Ethyl 3-hydroxyhexanoate

≥98%, FG

Synonym(s):

3-hydroxyhexanoic acid ethyl ester, Ethyl 3-hydroxy caproate

Sign Into View Organizational & Contract Pricing

Select a Size

W354503-SAMPLE-K
€47.40
100 G
€128.00
1 KG
€428.00
5 KG
€1,990.00

€47.40


Check Cart for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
W354503-SAMPLE-K
€47.40
100 G
€128.00
1 KG
€428.00
5 KG
€1,990.00

About This Item

Linear Formula:
CH3CH2CH2CH(OH)CH2CO2C2H5
CAS Number:
Molecular Weight:
160.21
FEMA Number:
3545
EC Number:
Council of Europe no.:
4101
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.535
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

€47.40


Check Cart for Availability

Request a Bulk OrderRequest more information

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002

Assay

≥98%

refractive index

n20/D 1.428 (lit.)

bp

90-92 °C/14 mmHg (lit.)

density

0.974 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

grape; green; citrus; fruity; sweet

SMILES string

CCCC(O)CC(=O)OCC

InChI

1S/C8H16O3/c1-3-5-7(9)6-8(10)11-4-2/h7,9H,3-6H2,1-2H3

InChI key

LYRIITRHDCNUHV-UHFFFAOYSA-N

Related Categories

General description

Ethyl 3-hydroxyhexanoate is the key volatile flavor compound of orange juice.[1] It also occurs in pineapple[2], wood apple[3], caja fruit[4] and tamarillo fruit.[5]

Application


  • Novel Antiviral Activity of Ethyl 3-Hydroxyhexanoate Against Coxsackievirus B Infection.: Discusses the potential antiviral properties of Ethyl 3-hydroxyhexanoate, positioning it as a candidate for therapeutic use against Coxsackievirus, with implications for pharmaceutical applications and virology (Olasunkanmi et al., 2022).

  • RIFM fragrance ingredient safety assessment, ethyl 3-hydroxyhexanoate, CAS Registry Number 2305-25-1.: A comprehensive safety assessment of Ethyl 3-hydroxyhexanoate used in fragrance formulations, emphasizing its toxicological profile and safe usage in consumer products (Api et al., 2020).


Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

201.2 °F - closed cup

Flash Point(C)

94 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of fermentation on free and bound volatile compounds of orange juice.
Fan G, et al.
Flavour and Fragrance Journal, 24(5), 219-225 (2009)
Aroma profiles of pineapple fruit (Ananas comosus [L.] Merr.) and pineapple products.
Elss S, et al.
LWT--Food Science and Technology, 38(3), 263-274 (2005)
Identification of volatile components of caja fruit (Spondias lutea L.) and chiral analysis of 3?hydroxy aliphatic esters.
Allegrone G & Barbeni M.
Flavour and Fragrance Journal, 7(6), 337-342 (1992)
Volatile constituents from Tamarillo (Cyphomandra betacea Sendtn.) fruit.
Torrado A, et al.
Flavour and Fragrance Journal, 10(6), 349-354 (1995)
Volatile flavor components of wood apple (Feronia limonia) and a processed product.
MacLeod AJ & Pieris NM.
Journal of Agricultural and Food Chemistry, 29(1), 49-53 (1981)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service