Skip to Content
Merck
All Photos(1)

Documents

W352117

Sigma-Aldrich

1-Propanethiol

natural, ≥98%, FG

Synonym(s):

Propyl mercaptan, n-Propylmercaptan, Mercaptan C3

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2SH
CAS Number:
Molecular Weight:
76.16
FEMA Number:
3521
Beilstein:
1696860
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
12.071

grade

FG
Halal
Kosher
natural

reg. compliance

EU Regulation 1334/2008 & 178/2002

vapor density

2.54 (vs air)

vapor pressure

122 mmHg ( 20 °C)

Assay

≥98%

refractive index

n20/D 1.426 (lit.)

bp

67-68 °C (lit.)

mp

−113 °C (lit.)

density

0.820 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

cabbage; onion; alliaceous

SMILES string

CCCS

InChI

1S/C3H8S/c1-2-3-4/h4H,2-3H2,1H3

InChI key

SUVIGLJNEAMWEG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Flam. Liq. 2 - Skin Sens. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-4.0 °F - closed cup

Flash Point(C)

-20 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service