Skip to Content
Merck
All Photos(1)

Documents

W340210

Sigma-Aldrich

cis-3-Hexenyl butyrate

natural, ≥95%, FG

Synonym(s):

(Z)-3-hexen-1-yl butyrate, Leaf butyrate, [(Z)-hex-3-enyl] butanoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2CO2CH2CH2CH=CHC2H5
CAS Number:
Molecular Weight:
170.25
FEMA Number:
3402
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.270
NACRES:
NA.21

grade

FG
Kosher
natural

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117

Assay

≥95%

refractive index

n20/D 1.431 (lit.)

bp

96 °C/20 mmHg (lit.)

density

0.885 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

fresh; green; vegetable; fruity

SMILES string

[H]\C(CC)=C(/[H])CCOC(=O)CCC

InChI

1S/C10H18O2/c1-3-5-6-7-9-12-10(11)8-4-2/h5-6H,3-4,7-9H2,1-2H3/b6-5-

InChI key

ZCHOPXVYTWUHDS-WAYWQWQTSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

174.2 °F - closed cup

Flash Point(C)

79 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lorenzo Boggia et al.
BMC plant biology, 15, 102-102 (2015-04-19)
The dynamics of plant volatile (PV) emission, and the relationship between damaged area and biosynthesis of bioactive molecules in plant-insect interactions, remain open questions. Direct Contact-Sorptive Tape Extraction (DC-STE) is a sorption sampling technique employing non adhesive polydimethylsiloxane tapes, which

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service