Skip to Content
Merck
All Photos(1)

Key Documents

W338303

Sigma-Aldrich

2-Pentylpyridine

≥97%, FG

Synonym(s):

2-Amyl pyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H15N
CAS Number:
Molecular Weight:
149.23
FEMA Number:
3383
EC Number:
Council of Europe no.:
11412
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
14.06
NACRES:
NA.21
grade:
FG
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

grade

FG

reg. compliance

EU Regulation 1334/2008 & 872/2012
FDA 21 CFR 110

Assay

≥97%

refractive index

n20/D 1.488 (lit.)

bp

102-107 °C (lit.)

density

0.897 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

fatty; green; herbaceous

SMILES string

CCCCCc1ccccn1

InChI

1S/C10H15N/c1-2-3-4-7-10-8-5-6-9-11-10/h5-6,8-9H,2-4,7H2,1H3

InChI key

HSDXVAOHEOSTFZ-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

174.2 °F - closed cup

Flash Point(C)

79 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yeşim Karademir et al.
Food research international (Ottawa, Ont.), 121, 919-925 (2019-05-22)
This study aimed to investigate lipid derived formations of decadien-1-amine, 2-pentylpyridine, and acrylamide in potato chips during frying. 2,4-Decadienal, a lipid derived carbonyl, was monitored in repeatedly used sunflower oil at different thermoxidation levels (0, 6, 12, 18, 24 h at

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service