Skip to Content
Merck
All Photos(1)

Key Documents

W338206

Sigma-Aldrich

Ethyl vinyl ketone

≥97%, stabilized with BHT, FG

Synonym(s):

1-Penten-3-one, Ethyl vinyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5COCH=CH2
CAS Number:
Molecular Weight:
84.12
FEMA Number:
3382
Beilstein:
1735857
EC Number:
Council of Europe no.:
11179
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
7.102
NACRES:
NA.21
Organoleptic:
garlic; onion; pungent
grade:
FG
Fragrance grade
Halal
Kosher
biological source:
synthetic
Agency:
follows IFRA guidelines
meets purity specifications of JECFA
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002

Assay

≥97%

contains

BHT as stabilizer

refractive index

n20/D 1.419 (lit.)

bp

38 °C/60 mmHg (lit.)

density

0.851 g/mL at 20 °C
0.845 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

garlic; onion; pungent

SMILES string

CCC(=O)C=C

InChI

1S/C5H8O/c1-3-5(6)4-2/h3H,1,4H2,2H3

InChI key

JLIDVCMBCGBIEY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethyl vinyl ketone is a volatile flavor compound mainly found in tomatoes.[1] It may be responsible for the raw bean odor and flavor in soybeans.[2]

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

14.0 °F - closed cup

Flash Point(C)

-10 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Identification of a volatile component in soybeans that contributes to the raw bean flavor.
Mattick LR & Hand DB.
Journal of Agricultural and Food Chemistry, 17(1),, 15-17 (1969)
Eduardo C Meurer et al.
Journal of mass spectrometry : JMS, 37(2), 146-154 (2002-02-22)
Gas-phase reactions of acylium ions with alpha,beta-unsaturated carbonyl compounds were investigated using pentaquadrupole multiple-stage mass spectrometry. With acrolein and metacrolein, CH(3)-C(+)(double bond)O, CH(2)(double bond)CH-C(+)(double bond)O, C(6)H(5)-C(+)(double bond)O, and (CH(3))(2)N-C(+)(double bond)O react to variable extents by mono and double polar [4
Weijun Liu et al.
Journal of the American Chemical Society, 128(5), 1426-1427 (2006-02-02)
The cycloaddition between 1,3-cyclohexadiene and various enones and enals (methyl vinyl ketone, ethyl vinyl ketone, methacrolien) is accomplished at room temperature in yields ranging from 51 to 68% without the use of Lewis acids, high pressures, or microwave reactors. This
E Eder et al.
Mutation research, 470(1), 29-37 (2000-09-15)
alpha,beta-Unsaturated ketones are bifunctional compounds which form promutagenic 1,N(2)-propanodeoxyguanosine adducts like carcinogenic alpha,beta-unsaturated aldehydes and are mutagenic and genotoxic like these aldehydes. They are important industrial chemicals, are found in our environment and are widespread in our food. We investigated
V Kettmann et al.
Acta crystallographica. Section C, Crystal structure communications, 57(Pt 6), 737-739 (2001-06-16)
The title compound, C18H19N3O2, was obtained by an azo-coupling reaction with enaminones and is composed of a planar azoenamine skeleton which forms a six-membered ring through a symmetrical intramolecular hydrogen bond. The compound was found to exist as an equilibrium

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service