Skip to Content
Merck
All Photos(1)

Documents

W337330

Sigma-Aldrich

2-Methyltetrahydrofuran-3-one

natural, 97%, FG

Synonym(s):

4,5-Dihydro-2-methyl-3(2H)-furanone, Coffee furanone, 2-Methyltetrahydro-3-furanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H8O2
CAS Number:
Molecular Weight:
100.12
FEMA Number:
3373
Beilstein:
1341334
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
13.042
NACRES:
NA.21

grade

FG
Halal
Kosher
natural

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002

Assay

97%

refractive index

n20/D 1.429 (lit.)

bp

139 °C (lit.)

density

1.034 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

coffee; nutty; brown; rum

SMILES string

CC1OCCC1=O

InChI

1S/C5H8O2/c1-4-5(6)2-3-7-4/h4H,2-3H2,1H3

InChI key

FCWYQRVIQDNGBI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

102.2 °F - closed cup

Flash Point(C)

39 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service