Skip to Content
Merck
All Photos(1)

Key Documents

W331104

Sigma-Aldrich

Methyl 2-thiofuroate

≥98%, FG

Synonym(s):

S-Methyl 2-thiofuroate

Sign Into View Organizational & Contract Pricing

Select a Size

W331104-SAMPLE-K
€48.80
25 G
€129.00
100 G
€224.00
250 G
€410.00

€48.80


Check Cart for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
W331104-SAMPLE-K
€48.80
25 G
€129.00
100 G
€224.00
250 G
€410.00

About This Item

Empirical Formula (Hill Notation):
C6H6O2S
CAS Number:
Molecular Weight:
142.18
FEMA Number:
3311
EC Number:
Council of Europe no.:
11547
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
13.142
NACRES:
NA.21
grade:
FG
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

€48.80


Check Cart for Availability

Request a Bulk OrderRequest more information

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002

Assay

≥98%

refractive index

n20/D 1.569 (lit.)

bp

63 °C/2 mmHg (lit.)

density

1.236 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

onion

SMILES string

CSC(=O)c1ccco1

InChI

1S/C6H6O2S/c1-9-6(7)5-3-2-4-8-5/h2-4H,1H3

InChI key

ISKUAGFDTRLBHG-UHFFFAOYSA-N

General description

Methyl 2-thiofuroate is a volatile sulfur compound reported to be formed during non-enzymatic browning reactions.[1][2]

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

201.2 °F - closed cup

Flash Point(C)

94.00 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lactose casein (Maillard) browning system: volatile components.
Ferretti A & Flanagan VP
Journal of Agricultural and Food Chemistry, 19(2), 245-249 (1971)
Sensory properties of volatile Maillard reaction products and related compounds: A literature review.
Fors S
ACS Symp. Ser., 215, 185?286-185?286 (1983)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service