Phytotherapy research : PTR, 20(1), 58-61 (2006-01-07)
The antifungal activity of the essential oil from Cinnamomum cassia, alone or combined with amphotericin B, a drug widely used for most indications despite side-effects was investigated. The composition of the oil was analysed by GC/MS and characterized by its
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica, 31(13), 1087-1090 (2006-10-20)
To observe the effect of 2-methoxycinnamaldehyde (isolated from fraction A of Guizhi Tang) on activity of COX and PGE2 release in rat cerebral microvascular endothelial cells (rCMEC) stimulated by IL-1. rCMEC were cultured, and identified by immunohistochemistry for von Willebrand
Prostaglandin E2 (PGE2) works as a common final mediator of the febrile. Guizhi-Tang, one of the most famous traditional Chinese medical formula used to treat influenza, common cold and other pyretic conditions, was previously reported to reduce the production of
Nuclear factor (NF)-cB is a transcription factor regulating the expression of inflammatory and immune genes. In the present study, an extract from stem bark of Cinnamomum cassia Blume(Lauraceae) was discovered to have an inhibitory effect on LPS-induced NF-KB transcriptional activity
Xenobiotica; the fate of foreign compounds in biological systems, 16(9), 845-852 (1986-09-01)
The metabolism of o-methoxycinnamaldehyde (1.3 mmol/kg, intragastrically) was studied in rats. Identification of the urinary metabolites by g.l.c.-mass spectrometry and quantification by h.p.l.c. showed that the major metabolic pathway (approx. two-thirds of the dose) was oxidation to the corresponding cinnamic