Skip to Content
Merck
All Photos(1)

Key Documents

W317705

Sigma-Aldrich

p-Mentha-8-thiol-3-one

mixture of cis and trans, ≥95%, FG

Synonym(s):

8-Mercaptomenthone, buchu mercaptan, 2-(1-Mercapto-1-methylethyl)-5-methylcyclohexanone

Sign Into View Organizational & Contract Pricing

Select a Size

W317705-SAMPLE-K
€47.40
100 G
€327.00
250 G
€584.00
1 KG
€1,470.00

€47.40


Check Cart for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
W317705-SAMPLE-K
€47.40
100 G
€327.00
250 G
€584.00
1 KG
€1,470.00

About This Item

Empirical Formula (Hill Notation):
C10H18OS
CAS Number:
Molecular Weight:
186.31
FEMA Number:
3177
EC Number:
Council of Europe no.:
11789
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
12.038
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

€47.40


Check Cart for Availability

Request a Bulk OrderRequest more information

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 110

Assay

≥95%

optical activity

[α]20/D −21°, neat

composition

may contain Pulegone

refractive index

n20/D 1.495 (lit.)

bp

56 °C/0.1 mmHg (lit.)

density

1 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

berry; minty; tropical; fruity; sulfurous

SMILES string

CC1CCC(C(=O)C1)C(C)(C)S

InChI

1S/C10H18OS/c1-7-4-5-8(9(11)6-7)10(2,3)12/h7-8,12H,4-6H2,1-3H3

InChI key

RVOKNSFEAOYULQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

p-Mentha-8-thiol-3-one can be used fragrance ingredient and also as a flavoring agent for its blackcurrant flavor.[1][2]

Application


  • RIFM fragrance ingredient safety assessment, p-mentha-8-thiol-3-one, CAS Registry Number 38462-22-5.: This review provides a comprehensive safety assessment of p-Mentha-8-thiol-3-one, focusing on its applications in fragrance formulations. The study evaluates the compound′s toxicological profile and discusses its safe usage levels in consumer products. The research highlights the compound′s potential as a fragrance ingredient due to its unique olfactory properties. (Api et al., 2021).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

226.4 °F - closed cup

Flash Point(C)

108 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electronic nose based discrimination of a perfumery compound in a fragrance.
Branca A, et al.
Sensors and Actuators B, Chemical, 92(1), 222-227 (2003)
Encyclopedia of Food and Color Additives, 3, 1137-1137 (2016)
Encyclopedia of Food and Color Additives, 3, 1137-1137 (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service