Skip to Content
Merck
All Photos(1)

Documents

W307300

Sigma-Aldrich

p-Tolyl acetate

≥98%, FCC

Synonym(s):

p-Cresyl acetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2C6H4CH3
CAS Number:
Molecular Weight:
150.17
FEMA Number:
3073
EC Number:
Council of Europe no.:
226
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.036
NACRES:
NA.21

biological source

synthetic

Agency

meets purity specifications of JECFA

reg. compliance

FCC
FDA 21 CFR 117
FDA 21 CFR 172.515

Assay

≥98%

refractive index

n20/D 1.501 (lit.)

bp

210-211 °C (lit.)

density

1.047 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

floral; animal; phenolic

SMILES string

CC(=O)Oc1ccc(C)cc1

InChI

1S/C9H10O2/c1-7-3-5-9(6-4-7)11-8(2)10/h3-6H,1-2H3

InChI key

CDJJKTLOZJAGIZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

194.0 °F - closed cup

Flash Point(C)

90 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service