Skip to Content
Merck
All Photos(1)

Key Documents

W304506

Sigma-Aldrich

Terpineol

mixture of isomers, 96%, FG

Synonym(s):

α,α,4-trimethyl-3-cyclohexene-1-methanol, 2-(4-methyl-3-cyclohexen-1-yl)-2-propanol, p-menth-1-en-8-ol

Sign Into View Organizational & Contract Pricing

Select a Size

W304506-SAMPLE-K
€47.40
1 KG
€67.90
10 KG
€260.00
25 KG
€478.00

€47.40


Check Cart for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
W304506-SAMPLE-K
€47.40
1 KG
€67.90
10 KG
€260.00
25 KG
€478.00

About This Item

Empirical Formula (Hill Notation):
C10H18O
CAS Number:
Molecular Weight:
154.25
FEMA Number:
3045
EC Number:
Council of Europe no.:
62
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
2.230
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

€47.40


Check Cart for Availability

Request a Bulk OrderRequest more information

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 172.515
FDA 21 CFR 178.1010

Assay

96%

form

viscous liquid

composition

d-terpineol, <1.0%
a-terpineol, ≥55.0%
b-terpineol, <10.0% (cis)
b-terpineol, <13.0% (trans)
g-terpineol, <23.0%

bp

213-218 °C (lit.)

solubility

ethanol: soluble 1.25ml/10ml, clear to slightly hazy, colorless to light yellow (50% ethanol)
ethanol: soluble 2.5ml/10ml, clear to slightly hazy, colorless to light yellow
ethanol: soluble 5mL/10mL, clear to slightly hazy, colorless to light yellow (70% Ethanol)

density

0.934 g/mL at 20 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

terpineol (mixture of isomers), α-terpineol

Organoleptic

lilac; woody; floral; citrus; pine

SMILES string

CC(=C)C1CCC(C)(O)CC1.CC2=CCC(CC2)C(C)(C)O

InChI

1S/2C10H18O/c1-8-4-6-9(7-5-8)10(2,3)11;1-8(2)9-4-6-10(3,11)7-5-9/h4,9,11H,5-7H2,1-3H3;9,11H,1,4-7H2,2-3H3

InChI key

MGABGZGUMNDCSN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

α-Terpineol is a cyclic monoterpene. It is one of the major monoterpenol component of the cultivar Gewürztraminer and other aroma-rich grape varieties.[1] α-Terpineol, a monoterpenoid alcohol, has been investigated for its anticonvulsant activity.[2] It was one of the insecticidal constituent of the essential oils of Dahlia pinnata.[3]FCC includes the following isomers in the listed purity: alpha-, (E)-beta, (Z)-beta, gamma-, terpinen-4-ol, and terpinen-1-ol.

Application

α-Terpineol is the suitable aroma standard used to study the influence of the drying method on volatile compounds of Origanum vulgare.[4] It may be employed as a highly viscous solvent in the preparation of photocatalytic transparent TiO2 thin film.[5]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

190.4 °F - Information taken from reference works and the literature.

Flash Point(C)

88 °C - Information taken from reference works and the literature.

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation of photocatalytic TiO2 transparent thin film by thermal decomposition of Ti-alkoxide with a-terpineol as a solvent.
Negishi N and Takeuchi K.
Thin Solid Films, 392(2), 249-253 (2001)
Evolution of the anticonvulsant activity of α-terpineol.
de Sousa DP, et al.
Pharmaceutical biology, 45(1), 69-70 (2007)
Diane M Martin et al.
Phytochemistry, 65(9), 1223-1229 (2004-06-09)
The flavour and aroma of certain Vitis vinifera grape varieties is dominated by volatile terpenes and small volatile aldehydes. Monoterpenes contribute to the final grape and wine aroma and flavour in form of free volatiles and as glycoside conjugates of
Francesca Mondello et al.
BMC infectious diseases, 6, 158-158 (2006-11-07)
Recent investigations on the antifungal properties of essential oil of Melaleuca alternifolia Cheel (Tea Tree Oil, TTO) have been performed with reference to the treatment of vaginal candidiasis. However, there is a lack of in vivo data supporting in vitro
Da-Cheng Wang et al.
Natural product research, 29(18), 1748-1751 (2015-01-08)
The aim of this research was to determine the chemical composition of the essential oils of Dahlia pinnata, their insecticidal activity against Sitophilus zeamais and Sitophilusoryzae and to isolate insecticidal constituents. Based on bioactivity-guided fractionation, active constituents were isolated and

Protocols

GC Analysis of Sweet Orange Essential Oil on SLB®-5ms (10 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service