Skip to Content
Merck
All Photos(1)

Key Documents

W303208

Sigma-Aldrich

Spearmint oil

rectified, FG

Synonym(s):

Mentha spicata

Sign Into View Organizational & Contract Pricing

Select a Size

W303208-SAMPLE-K
€49.60
1 KG
€195.00
4 KG
€730.00
9 KG
€1,270.00

€49.60


Check Cart for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
W303208-SAMPLE-K
€49.60
1 KG
€195.00
4 KG
€730.00
9 KG
€1,270.00

About This Item

CAS Number:
FEMA Number:
3032
MDL number:
UNSPSC Code:
12164502
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
biological source:
(Mentha cardiaca)
food allergen:
no known allergens

€49.60


Check Cart for Availability

Request a Bulk OrderRequest more information

biological source

(Mentha cardiaca)

Quality Level

300
400

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 172.230
FDA 21 CFR 182.20

optical activity

[α]20/D −54°, neat

composition

Contains IFRA restricted Carvone and EU 1223/2009 restricted Limonene

origin

USA origin

refractive index

n20/D 1.489 (lit.)

bp

228 °C (lit.)

density

0.918 g/mL at 25 °C

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

Mentha spicata, α-pinene and β-pinene, limonene (sum of D, L and DL), carvone

Organoleptic

spearmint

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application


  • Effects of ingested essential oils and propolis extracts on honey bee (Hymenoptera: Apidae) health and gut microbiota.: This research highlights the potential of spearmint oil to improve the health and survival of honey bees, addressing challenges like pathogen susceptibility and gut microbiota balance, essential for sustaining bee populations and by extension, global agriculture (Martin Ewert A et al., 2023).

Biochem/physiol Actions

Taste at 10 ppm

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

201.2 °F - closed cup

Flash Point(C)

94 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Mosciano, G.
Perfumer & Flavorist, 27, 92-92 (2002)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service