Skip to Content
Merck
All Photos(1)

Key Documents

W302475

Sigma-Aldrich

Sodium acetate

anhydrous, natural (US), ≥99%, FG

Synonym(s):

Acetic acid sodium salt

Sign Into View Organizational & Contract Pricing

Select a Size

W302475-SAMPLE-K
€49.00
1 KG
€98.30
10 KG
€294.00

€49.00


Estimated to ship on01 April 2025


Request a Bulk OrderRequest more information

Select a Size

Change View
W302475-SAMPLE-K
€49.00
1 KG
€98.30
10 KG
€294.00

About This Item

Linear Formula:
CH3COONa
CAS Number:
Molecular Weight:
82.03
FEMA Number:
3024
Beilstein:
3595639
MDL number:
UNSPSC Code:
12164502
E Number:
E 262(i)
NACRES:
NA.21
Organoleptic:
odorless
grade:
FG
Fragrance grade
Halal
Kosher
anhydrous
natural (US)
biological source:
botanical
Agency:
follows IFRA guidelines
meets purity specifications of JECFA
food allergen:
no known allergens

€49.00


Estimated to ship on01 April 2025


Request a Bulk OrderRequest more information

biological source

botanical

Quality Level

grade

FG
Fragrance grade
Halal
Kosher
anhydrous
natural (US)

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1333/2008 & 178/2002
FDA 21 CFR 117

Assay

≥99%

form

solid

autoignition temp.

1112 °F

pKa 

4.76 (acetic acid)

mp

>300 °C (dec.) (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

odorless

SMILES string

[Na+].CC([O-])=O

InChI

1S/C2H4O2.Na/c1-2(3)4;/h1H3,(H,3,4);/q;+1/p-1

InChI key

VMHLLURERBWHNL-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.
Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service