Skip to Content
Merck
All Photos(1)

Documents

W300403

Sigma-Aldrich

Salicylaldehyde

≥98%, FG

Synonym(s):

2-Hydroxybenzaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2-(HO)C6H4CHO
CAS Number:
Molecular Weight:
122.12
FEMA Number:
3004
Beilstein:
471388
Council of Europe no.:
605
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
5.055
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 872/2012
FDA 21 CFR 172.515

vapor density

4.2 (vs air)

vapor pressure

1 mmHg ( 33 °C)

Assay

≥98%

refractive index

n20/D 1.573 (lit.)

bp

197 °C (lit.)

mp

1-2 °C (lit.)

density

1.146 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

salicylaldehyde

Organoleptic

medicinal; cooling; spicy

SMILES string

Oc1ccccc1C=O

InChI

1S/C7H6O2/c8-5-6-3-1-2-4-7(6)9/h1-5,9H

InChI key

SMQUZDBALVYZAC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Salicylaldehyde is the main odor constituent of buckwheat flour. It is also found in anise and vanilla extracts.

Application

  • Facile synthesis of iminated lignin for enhanced free radical and lead ion scavenging capabilities.: This study presents a novel method for synthesizing iminated lignin using salicylaldehyde. The modified lignin demonstrates significantly improved free radical and lead ion scavenging abilities, highlighting its potential application in environmental remediation and biochemical processes (Xia et al., 2024).

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

170.6 °F - closed cup

Flash Point(C)

77 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Salicylaldehyde is a characteristic aroma component of buckwheat groats.
Janes D & Kreft S.
Food Chemistry, 109(2), 293-298 (2008)
Spectrophotometric profiles of off-flavor aldehydes by using their reactions with 2-thiobarbituric acid.
Guzman-Chozas M, et al.
Journal of Agricultural and Food Chemistry, 45(7), 2452-2457 (1997)
Rhodium(III)-catalyzed dehydrogenative Heck reaction of salicylaldehydes.
Zhuangzhi Shi et al.
Angewandte Chemie (International ed. in English), 51(32), 8092-8096 (2012-06-30)
Ziyad A Taha et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 81(1), 317-323 (2011-07-15)
Eight new lanthanide metal complexes [LnL(NO(3))(2)]NO(3) {Ln(III) = Nd, Dy, Sm, Pr, Gd, Tb, La and Er, L = bis-(salicyladehyde)-1,3-propylenediimine Schiff base ligand} were prepared. These complexes were characterized by elemental analysis, thermogravimetric analysis (TGA), molar conductivity measurements and spectral
Wan Yen Lee et al.
Metallomics : integrated biometal science, 4(2), 188-196 (2011-12-03)
The copper(ii) complexes of two salicylaldehyde semicarbazones, HOC(6)H(4)CH[double bond, length as m-dash]N-NHCONR(2) [H(2)Bnz(2) (R = CH(2)Ph) and H(2)Bu(2) (R = Bu)], were evaluated for their DNA binding and cleavage properties by spectrophotometric DNA titration, ethidium bromide displacement assay and electrophoretic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service