Skip to Content
Merck
All Photos(1)

Key Documents

W296000

Sigma-Aldrich

Propyl isovalerate

natural (US), ≥98%, FG

Synonym(s):

Propyl 3-methylbutanoate, Propyl 3-methylbutyrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2COO(CH2)2CH3
CAS Number:
Molecular Weight:
144.21
FEMA Number:
2960
Beilstein:
1747146
EC Number:
Council of Europe no.:
443
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.448
NACRES:
NA.21
Pricing and availability is not currently available.

grade

FG
Fragrance grade
Halal
Kosher
natural (US)

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 872/2012

Assay

≥98%

density

0.862 g/mL at 20 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

apple; fruity; sweet

SMILES string

CCCOC(=O)CC(C)C

InChI

1S/C8H16O2/c1-4-5-10-8(9)6-7(2)3/h7H,4-6H2,1-3H3

InChI key

LSJMDWFAADPNAX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Natural occurence: Apple, banana, gruyere cheese and jackfruit [1]

Application

The non-specific fruitiness of this compound makes it a good fit for many types of fruit flavors, including apple, banana, blueberry, pear, peach, grape, strawberry, pineapple and tropicals like jackfruit and durian. These same notes will also add to the fruity complex of aged cheese like Parmesan, Asiago, and Romano.[1]

Biochem/physiol Actions

Odor at 1%[1]
Taste at 5-10ppm[1]

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

111.2 °F

Flash Point(C)

44 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Organoleptic Characteristics of Flavor Materials
Michalski, J.
Perfumer & Flavorist, 39(4), 50-50 (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service