Skip to Content
Merck
All Photos(1)

Documents

W276618

Sigma-Aldrich

Myrrh oil

Synonym(s):

Commiphora molmol spp.

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
FEMA Number:
2766
MDL number:
UNSPSC Code:
12164502

biological source

Commiphora spp.

optical activity

[α]20/D −92°, neat

origin

France origin

refractive index

n20/D 1.52 (lit.)

bp

220 °C (lit.)

density

1.003 g/mL at 25 °C (lit.)

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Integrating complementary and alternative medicine: use of myrrh in wound management.
M Eileen Walsh et al.
Journal of vascular nursing : official publication of the Society for Peripheral Vascular Nursing, 28(3), 102-102 (2010-08-17)
Jacob George et al.
Indian journal of dental research : official publication of Indian Society for Dental Research, 20(4), 480-482 (2010-02-09)
A double-blinded controlled clinical trial with parallel groups was designed to investigate the effectiveness of a herbal-based toothpaste in the control of plaque and gingivitis as compared with a conventional dentifrice. The efficacy of Colgate Herbal over Colgate tooth paste
Mostafa Yakoot
Arquivos de gastroenterologia, 47(4), 393-394 (2011-01-13)
Mirazid® is a patented preparation from a plant that had been used in folk medicine since ancient Egyptians (Myrrh). It was registered in Egypt for the treatment of schistosomiasis and fascioliasis. Over 32 independent studies for efficacy of Mirazid had
Xiao-ling Wang et al.
Acta pharmacologica Sinica, 32(3), 338-344 (2011-03-05)
To examine whether two naturally occurring sesquiterpenoids (ST1 and ST2) with anti-proliferative activity in prostate cancer cells inhibit androgen receptor (AR) signaling. Human prostate cancer cell lines LNCaP and PC3 were used. The expression of AR, AR translocation into the
Feng Shi et al.
International journal of nanomedicine, 7, 2033-2043 (2012-05-24)
The aim of the present study was to prepare solid lipid nanoparticles (SLNs) for the oral delivery of frankincense and myrrh essential oils (FMO). Aqueous dispersions of SLNs were successfully prepared by a high-pressure homogenization method using Compritol 888 ATO

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service