Skip to Content
Merck
All Photos(1)

Key Documents

W274003

Sigma-Aldrich

4-Methyl-1-phenyl-2-pentanone

≥95%, FG

Synonym(s):

Benzyl isobutyl ketone

Sign Into View Organizational & Contract Pricing

Select a Size

W274003-SAMPLE-K
€49.30
100 G
€498.00

€49.30


Estimated to ship on31 May 2025


Request a Bulk OrderRequest more information

Select a Size

Change View
W274003-SAMPLE-K
€49.30
100 G
€498.00

About This Item

Linear Formula:
(CH3)2CHCH2COCH2C6H5
CAS Number:
Molecular Weight:
176.25
FEMA Number:
2740
EC Number:
Council of Europe no.:
159
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
7.025
NACRES:
NA.21
grade:
FG
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

€49.30


Estimated to ship on31 May 2025


Request a Bulk OrderRequest more information

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 172.515

Assay

≥95%

impurities

1.6% 4-methyl-1-phenyl-2-pentanol

refractive index

n20/D 1.498 (lit.)

bp

250-251 °C (lit.)

density

0.949 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

woody; spicy; sweet

storage temp.

2-8°C

SMILES string

CC(C)CC(=O)Cc1ccccc1

InChI

1S/C12H16O/c1-10(2)8-12(13)9-11-6-4-3-5-7-11/h3-7,10H,8-9H2,1-2H3

InChI key

DTYGTEGDVPAKDA-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

221.0 °F - closed cup

Flash Point(C)

105.00 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service