Skip to Content
Merck
All Photos(3)

Key Documents

W271810

Sigma-Aldrich

Dimethyl anthranilate

greener alternative

natural, ≥98%, FG

Synonym(s):

Methyl N-methylanthranilate

Sign Into View Organizational & Contract Pricing

Select a Size

W271810-SAMPLE-K
€49.30
100 G
€147.00
1 KG
€882.00

€49.30


Check Cart for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
W271810-SAMPLE-K
€49.30
100 G
€147.00
1 KG
€882.00

About This Item

Linear Formula:
2-(CH3NH)C6H4CO2CH3
CAS Number:
Molecular Weight:
165.19
FEMA Number:
2718
Beilstein:
607217
EC Number:
Council of Europe no.:
756c
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.781
NACRES:
NA.21
grade:
FG
Kosher
natural
food allergen:
no known allergens

€49.30


Check Cart for Availability

Request a Bulk OrderRequest more information

grade

FG
Kosher
natural

Quality Level

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 172.515

Assay

≥98%

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

refractive index

n20/D 1.579 (lit.)

bp

256 °C (lit.)

mp

17-19 °C (lit.)

density

1.126 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

greener alternative category

Organoleptic

grape skin; fruity; floral; sweet

SMILES string

CNc1ccccc1C(=O)OC

InChI

1S/C9H11NO2/c1-10-8-6-4-3-5-7(8)9(11)12-2/h3-6,10H,1-2H3

InChI key

GVOWHGSUZUUUDR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dimethyl anthranilate is one of the main volatile constituents in citrus species such as mandarin[1], and calamondin.[2]

Application


  • Effects of Dimethyl Anthranilate-Based Repellents on Behavior, Plumage Condition, Egg Quality, and Performance in Laying Hens.: Investigates the impact of Dimethyl Anthranilate-based repellents on laying hens, focusing on their behavior, plumage condition, egg quality, and overall performance (Terčič et al., 2020).

  • A simple method to discriminate Guangchenpi and Chenpi by high-performance thin-layer chromatography and high-performance liquid chromatography based on analysis of dimethyl anthranilate.: Presents a chromatographic method to differentiate between Guangchenpi and Chenpi, emphasizing the role of Dimethyl Anthranilate in this differentiation process (Li et al., 2019).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

216.7 °F - closed cup

Flash Point(C)

102.6 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Field evaluation of dimethyl anthranilate as a bird repellent livestock feed additive.
Mason JR, et al.
The Journal of wildlife management, 636-642 (1985)
Analytical characterization of mandarin (Citrus deliciosa Ten.) essential oil.
Dugo P, et al.
Flavour and Fragrance Journal, 26(1), 34-46 (2011)
Dimethyl anthranilate as a bird repellent in livestock feed.
Glahn JF, et al.
Wildlife Society Bulletin, 17(3), 313-320 (1989)
Volatile constituents of calamondin peel and juice (Citrus madurensis Lour.) cultivated in the Philippines.
Takeuchi H, et al.
J. Essent. Oil Res., 17(1), 23-26 (2005)
Hayato Nishida et al.
Regulatory toxicology and pharmacology : RTP, 72(3), 578-585 (2015-06-07)
Previously, a non-animal screening approach was proposed for evaluating photosafety of cosmetic ingredients by means of in vitro photochemical and photobiochemical assays; however, complex cosmetic ingredients, such as plant extracts and polymers, could not be evaluated because their molecular weight

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service