Skip to Content
Merck
All Photos(1)

Key Documents

W257605

Sigma-Aldrich

Hexyl propionate

≥97%, FG

Sign Into View Organizational & Contract Pricing

Select a Size

W257605-SAMPLE-K
€47.40
1 KG
€95.70
20 KG
€809.00

€47.40


Check Cart for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
W257605-SAMPLE-K
€47.40
1 KG
€95.70
20 KG
€809.00

About This Item

Linear Formula:
C2H5CO2(CH2)5CH3
CAS Number:
Molecular Weight:
158.24
FEMA Number:
2576
EC Number:
Council of Europe no.:
420
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.139
NACRES:
NA.21
grade:
FG
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

€47.40


Check Cart for Availability

Request a Bulk OrderRequest more information

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
FDA 21 CFR 172.515

Assay

≥97%

refractive index

n20/D 1.413 (lit.)

bp

73-74 °C/10 mmHg (lit.)

density

0.871 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

green; musty; fruity; pear

storage temp.

2-8°C

SMILES string

CCCCCCOC(=O)CC

InChI

1S/C9H18O2/c1-3-5-6-7-8-11-9(10)4-2/h3-8H2,1-2H3

InChI key

GOKKOFHHJFGZHW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Taste at 10 ppm

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

149.0 °F - closed cup

Flash Point(C)

65 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Organoleptic Characteristics of Flavor Materials
Mosciano, G.
Perfumer & Flavorist, 28, 77-77 (2003)
Rachel S Leisso et al.
Physiologia plantarum, 153(2), 204-220 (2014-06-20)
'Soggy breakdown' (SB) is an internal flesh disorder of 'Honeycrisp' apple (Malus × domestica Borkh.) fruit that occurs during low temperature storage. The disorder is a chilling injury (CI) in which visible symptoms typically appear after several weeks of storage

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service