Skip to Content
Merck
All Photos(1)

Key Documents

W247618

Sigma-Aldrich

Methyl isoeugenol

≥98%, FG

Synonym(s):

1,2-Dimethoxy-4-propenylbenzene, Isoeugenyl methyl ether

Sign Into View Organizational & Contract Pricing

Select a Size

W247618-SAMPLE-K
€47.40
250 G
€83.60
1 KG
€132.00
10 KG
€1,040.00

€47.40


Check Cart for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
W247618-SAMPLE-K
€47.40
250 G
€83.60
1 KG
€132.00
10 KG
€1,040.00

About This Item

Linear Formula:
CH3CH=CHC6H3(OCH3)2
CAS Number:
Molecular Weight:
178.23
FEMA Number:
2476
EC Number:
Council of Europe no.:
186
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
4.013
NACRES:
NA.21
grade:
FG
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

€47.40


Check Cart for Availability

Request a Bulk OrderRequest more information

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
FDA 21 CFR 172.515

Assay

≥98%

refractive index

n20/D 1.568 (lit.)

bp

262-264 °C (lit.)

mp

98-100 °C (lit.)

density

1.05 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

spicy

SMILES string

COc1ccc(\C=C\C)cc1OC

InChI

1S/C11H14O2/c1-4-5-9-6-7-10(12-2)11(8-9)13-3/h4-8H,1-3H3/b5-4+

InChI key

NNWHUJCUHAELCL-SNAWJCMRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methyl isoeugenol is one of the main components found in the essential oil extracted from Acorus calamus[1], bark of Croton malambo[2] and rhizomes of Zingiber zerumbet, Hedychium coronarium and Etlingera cevuga.[3]

Application

Methyl isoeugenol can be used as a natural food flavor in food industries.[4][5]

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anxiolytic and antidepressant like effects of natural food flavour (E)-methyl isoeugenol
Fajemiroye JO, et al.
Food & Function, 5(8), 1819-1828 (2014)
Hypotensive and vasorelaxant effects of (E)-Methyl isoeugenol: A naturally occurring food flavour
Fajemiroye JO, et al.
Food And Chemical Toxicology, 70, 214-221 (2014)
Major constituents and antimicrobial activity of Korean herb Acorus calamus.
Kim WJ, et al.
Natural product research, 25(13), 1278-1281 (2011)
Aromatic plants of French Polynesia. I. Constituents of the essential oils of rhizomes of three Zingiberaceae: Zingiber zerumbet Smith, Hedychium coronarium Koenig and Etlingera cevuga Smith.
Lechat-Vahirua I, et al.
J. Essent. Oil Res., 5(1), 55-59 (1993)
[In vitro study of the expectorant effect of phenylpropane derivatives of hazelwort. 12. Active substances form Asarum europaeum. L].
L Gracza
Planta medica, 42(2), 155-159 (1981-06-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service