Skip to Content
Merck
All Photos(1)

Key Documents

W243507

Sigma-Aldrich

Ethyl 3-(furan-2-yl)propionate

≥98%, FG

Synonym(s):

Ethyl 3-(2-furyl)propanoate

Sign Into View Organizational & Contract Pricing

Select a Size

W243507-SAMPLE-K
€60.30
100 G
€249.00
1 KG
€2,270.00

€60.30


Check Cart for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
W243507-SAMPLE-K
€60.30
100 G
€249.00
1 KG
€2,270.00

About This Item

Empirical Formula (Hill Notation):
C9H12O3
CAS Number:
Molecular Weight:
168.19
FEMA Number:
2435
EC Number:
Council of Europe no.:
2091
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
13.022
NACRES:
NA.21
grade:
FG
Kosher
biological source:
synthetic
food allergen:
no known allergens

€60.30


Check Cart for Availability

Request a Bulk OrderRequest more information

biological source

synthetic

Quality Level

grade

FG
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
FDA 21 CFR 172.515

Assay

≥98%

refractive index

n20/D 1.459 (lit.)

bp

212 °C (lit.)

density

1.054 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

fruity; pineapple; sweet

SMILES string

CCOC(=O)CCc1ccco1

InChI

1S/C9H12O3/c1-2-11-9(10)6-5-8-4-3-7-12-8/h3-4,7H,2,5-6H2,1H3

InChI key

OWIWZQQFSTZZIG-UHFFFAOYSA-N

General description

Ethyl 3-(furan-2-yl)propionate can be used as a flavoring agent in the food industry.[1]

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

195.8 °F - closed cup

Flash Point(C)

91 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Burdock, GA
Encyclopedia of Food and Color Additives, 1, 985-986 (1997)
Burdock, GA
Encyclopedia of Food and Color Additives, 1, 985-986 (1997)
Cristina Avonto et al.
Toxicology and applied pharmacology, 318, 16-22 (2017-01-23)
German chamomile is one of the most popular herbal ingredients used in cosmetics and personal care products. Allergic skin reactions following topical application of German chamomile have been occasionally reported, although it is not fully understood which of the chemical

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service