Skip to Content
Merck
All Photos(1)

Key Documents

W226318

Sigma-Aldrich

Castor oil

Certified organic (NOP)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
FEMA Number:
2263
EC Number:
MDL number:
UNSPSC Code:
12164502
grade:
Certified organic (NOP)
Halal
biological source:
Ricinus communis
food allergen:
no known allergens
Pricing and availability is not currently available.

biological source

Ricinus communis

grade

Certified organic (NOP)
Halal

reg. compliance

FDA 21 CFR 117
FDA 21 CFR 182.20

origin

India origin

refractive index

n20/D 1.478 (lit.)

bp

313 °C (lit.)

density

0.955 g/mL at 25 °C

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

faint

Looking for similar products? Visit Product Comparison Guide

Legal Information

Certified organic by MOSA

Storage Class Code

12 - Non Combustible Liquids

WGK

awg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alejandro Ruiz Olivares et al.
Journal of environmental management, 114, 316-323 (2012-11-23)
Bioenergy production combined with phytoremediation has been suggested to help in solving two critical world problems: the gradual reduction of fossil fuels and soil contamination. The aim of this research was to investigate the potential for the use of Ricinus
Luiz Fernando Kubrusly et al.
Revista brasileira de cirurgia cardiovascular : orgao oficial da Sociedade Brasileira de Cirurgia Cardiovascular, 27(3), 392-400 (2013-01-05)
The aim of the present investigation is to determine if the tissue reaction to the Riccinus communis (mamona) polymer has significant statistical difference compared to the tissue reaction provoked by the titanium implant. Thirty two Cavia porcellus were divided into
E C Alonso et al.
Journal of economic entomology, 106(2), 742-746 (2013-06-22)
Leaf-cutting ants are the main herbivores in the New World tropics. Although the toxicity of seed oils against these ants has been poorly investigated, previous results revealed that seed oils exert considerable toxic activity against these insects. This paper analyzes
S Rajan et al.
Asian Pacific journal of tropical medicine, 5(8), 630-633 (2012-07-31)
To examine the antidiarrhoeal activity of alcoholic and aqueous seed kernel extract of Mangifera indica (M. indica) on castor oil-induced diarrhoeal activity in Swiss albino mice. Mango seed kernels were processed and extracted using alcohol and water. Antidiarrhoeal activity of
Marcia Carneiro Valera et al.
General dentistry, 60(4), e204-e209 (2012-07-12)
This in vitro study sought to evaluate the effectiveness of castor oil extract used as an irrigating solution on Escherichia coli and its endotoxins in root canals. Sixty single-rooted teeth were prepared (using castor oil extract as irrigating solution) and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service