Skip to Content
Merck
All Photos(1)

Key Documents

W219800

Sigma-Aldrich

Isobutyl 3-(2-furan)propionate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H16O3
CAS Number:
Molecular Weight:
196.24
FEMA Number:
2198
EC Number:
Council of Europe no.:
2093c
MDL number:
UNSPSC Code:
12164502
Flavis number:
13.024

biological source

synthetic

SMILES string

CC(C)COC(=O)CCc1ccco1

Application

Nut flavors, caramel, cherry, pineapple and pear, cinnamon nuances, various fruit blends, brandy and baked goods.

Features and Benefits

Odor: @ 1.0 percent. Sweet fruity, ripe pineapple, cherry, slightly cinnamon spicy, brown caramel and slightly balsamic.
Taste: @ 10 PPM. Winey, fruity pineapple, cherry and honey with slight spicy cinnamic and berry nuances.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Organoleptic Characteristics of Flavor Materials
Mosciano, G.
Perfumer & Flavorist, 30(5), 49-49 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service