Skip to Content
Merck
All Photos(1)

Key Documents

W213306

Sigma-Aldrich

Benzoin resin absolute, Siam

natural

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
FEMA Number:
2133
EC Number:
MDL number:
UNSPSC Code:
12164502
grade:
Kosher
natural
biological source:
gum benzoin, Siam
food allergen:
no known allergens

biological source

gum benzoin, Siam

Quality Level

grade

Kosher
natural

origin

France origin

mp

60-70 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

balsam

Preparation Note

Extraction method: solvent extraction and distillation

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service