Skip to Content
Merck
All Photos(1)

Key Documents

W206318

Sigma-Aldrich

Isoamyl cinnamate

≥97%, FG

Synonym(s):

Isopentyl cinnamate, Isoamyl 3-phenyl propenoate

Sign Into View Organizational & Contract Pricing

Select a Size

W206318-SAMPLE-K
€48.80
1 KG
€159.00
10 KG
€1,130.00

€48.80


Check Cart for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
W206318-SAMPLE-K
€48.80
1 KG
€159.00
10 KG
€1,130.00

About This Item

Linear Formula:
C6H5CH=CHCO2CH2CH2CH(CH3)2
CAS Number:
Molecular Weight:
218.29
FEMA Number:
2063
EC Number:
Council of Europe no.:
335
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.742
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

€48.80


Check Cart for Availability

Request a Bulk OrderRequest more information

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
FDA 21 CFR 172.515

Assay

≥97%

refractive index

n20/D 1.536 (lit.)

bp

310 °C (lit.)

density

0.995 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

cocoa; balsamic

SMILES string

CC(C)CCOC(=O)\C=C\c1ccccc1

InChI

1S/C14H18O2/c1-12(2)10-11-16-14(15)9-8-13-6-4-3-5-7-13/h3-9,12H,10-11H2,1-2H3/b9-8+

InChI key

JFHCDEYLWGVZMX-CMDGGOBGSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Isoamyl cinnamate is mainly used as a fragrance ingredient[1] and also as a food flavoring agent.[2]

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Burdock GA
Encyclopedia of Food and Color Additives, 1, 1422-1422 (1997)
Burdock GA
Encyclopedia of Food and Color Additives, 1, 1422-1422 (1997)
Fragrance material review on isoamyl cinnamate
Bhatia SP, et al.
Food And Chemical Toxicology, 45(1), S98-S101 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service