Skip to Content
Merck
All Photos(1)

Key Documents

W204307

Sigma-Aldrich

Allyl tiglate

≥97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH=C(CH3)CO2CH2CH=CH2
CAS Number:
Molecular Weight:
140.18
FEMA Number:
2043
EC Number:
Council of Europe no.:
2183
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.493

biological source

synthetic

reg. compliance

FDA 21 CFR 172.515

Assay

≥97%

refractive index

n20/D 1.453 (lit.)

density

0.926 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

berry; green; jam

SMILES string

C\C=C(/C)C(=O)OCC=C

InChI

1S/C8H12O2/c1-4-6-10-8(9)7(3)5-2/h4-5H,1,6H2,2-3H3/b7-5+

InChI key

ODOZNBUSHKFCSH-FNORWQNLSA-N

Packaging

Packaged in glass bottles

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

140.0 °F - closed cup

Flash Point(C)

60 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jenelle L Wallace et al.
Neuron, 96(4), 883-896 (2017-10-24)
New neurons appear only in a few regions of the adult mammalian brain and become integrated into existing circuits. Little is known about the functional development of individual neurons in vivo. We examined the functional life history of adult-born granule cells

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service