Skip to Content
Merck
All Photos(2)

Key Documents

V1805

Sigma-Aldrich

4-Vinylbiphenyl

Synonym(s):

4-Phenylstyrene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C6H4CH=CH2
CAS Number:
Molecular Weight:
180.25
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

mp

119-121 °C (lit.)

storage temp.

2-8°C

SMILES string

C=Cc1ccc(cc1)-c2ccccc2

InChI

1S/C14H12/c1-2-12-8-10-14(11-9-12)13-6-4-3-5-7-13/h2-11H,1H2

InChI key

HDBWAWNLGGMZRQ-UHFFFAOYSA-N

Hazard Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service