Key Documents
TEM-71166-SP
Sigma-Aldrich
Gripper Tool for Cryo Grid Boxes
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
General description
Use this tool to open the lid of our cryogrid box (TEM-71166-40). It features a spring loaded vise that grips the lid securely and a ribbed barrel for ease in unscrewing the lid. Made of high quality aluminum, it can be used with liquid nitrogen. Can also be used to carry the cryo grid box.