Skip to Content
Merck
All Photos(1)

Key Documents

T70408

Sigma-Aldrich

3,4,5-Trimethoxycinnamic acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)3C6H2CH=CHCO2H
CAS Number:
Molecular Weight:
238.24
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

mp

125-127 °C (lit.)

SMILES string

COc1cc(\C=C\C(O)=O)cc(OC)c1OC

InChI

1S/C12H14O5/c1-15-9-6-8(4-5-11(13)14)7-10(16-2)12(9)17-3/h4-7H,1-3H3,(H,13,14)/b5-4+

InChI key

YTFVRYKNXDADBI-SNAWJCMRSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Zhenghang Zhao et al.
Biological & pharmaceutical bulletin, 36(2), 238-244 (2012-12-01)
3,4,5-Trimethoxycinnamic acid (TMCA), methyl 3,4,5-trimethoxycinnamate (M-TMCA) and p-methoxycinnamic acid (PMCA) have been identified as the major bioactive components in the serum collected from rats treated with oral administration of Polygalae Radix ("YuanZhi," the roots of Polygala tenuifolia WILLD.), a traditional
Sarvesh Kumar et al.
Biochemistry, 44(48), 15944-15952 (2005-11-30)
We report here the isolation and characterization of two active principles, ethyl 3',4',5'-trimethoxycinnamate (1) and piperine (2), from the combined hexane and chloroform extracts of Piper longum. Using primary human umbilical vein endothelial cells, we evaluated the activities of compound
Yingming Wu et al.
Organic letters, 11(3), 597-600 (2008-12-25)
A highly flexible and concise total synthesis of (+)-podophyllotoxin featured with an enantioselective sequential conjugate addition-allylation reaction was reported. Starting from commercially available 3,4,5-trimethoxycinnamic acid, this new route leads to (+)-podophyllotoxin 1 in only eight steps with 29% overall yield.
Keiko Kawashima et al.
Biological & pharmaceutical bulletin, 27(8), 1317-1319 (2004-08-12)
3,4,5-trimethoxycinnamic acid (TMCA) is one of the constituents in Onji (roots of Polygala tenuifolia WILLD), an herbal medicine used for sedative in Japanese traditional Kampo medicine. Our previous study revealed that oral administration of this compound prolongs sleeping time induced
[The detection of Azotobacter and its significance in criminal technical soil examinations].
E Kissling
Archiv fur Kriminologie, 165(1-2), 27-34 (1980-01-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service