T5702
Tetrabutyl orthosilicate
97%
Synonym(s):
Tetrabutoxysilane
About This Item
Recommended Products
vapor density
>1 (vs air)
Assay
97%
form
liquid
refractive index
n20/D 1.413 (lit.)
bp
275 °C (lit.)
density
0.899 g/mL at 25 °C (lit.)
SMILES string
CCCCO[Si](OCCCC)(OCCCC)OCCCC
InChI
1S/C16H36O4Si/c1-5-9-13-17-21(18-14-10-6-2,19-15-11-7-3)20-16-12-8-4/h5-16H2,1-4H3
InChI key
UQMOLLPKNHFRAC-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Mesoporous materials, such as aerogels, offer advantages for practical hydrogen storage. They have large surface areas, open porosity, small pore sizes, and the ability to coat the surface with one or more compounds.
Silica's versatility spans various industries, including biomedical applications.
Hybrid organic-inorganic sol-gel materials containing silica were first called “ORMOSILs” in 1984.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service