Skip to Content
Merck
All Photos(1)

Key Documents

T27952

Sigma-Aldrich

1-(2-Thienyl)-1-propanone

98%

Synonym(s):

2-Propionylthiophene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H8OS
CAS Number:
Molecular Weight:
140.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.553 (lit.)

bp

107 °C/11 mmHg (lit.)

density

1.126 g/mL at 25 °C (lit.)

SMILES string

CCC(=O)c1cccs1

InChI

1S/C7H8OS/c1-2-6(8)7-4-3-5-9-7/h3-5H,2H2,1H3

InChI key

MFPZQZZWAMAHOY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

213.8 °F - closed cup

Flash Point(C)

101 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Robert J Cannon et al.
Journal of agricultural and food chemistry, 63(7), 1915-1931 (2015-02-03)
Lemons (Citrus limon) are a desirable citrus fruit grown and used globally in a wide range of applications. The main constituents of this sour-tasting fruit have been well quantitated and characterized. However, additional research is still necessary to better understand

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service